COM497518279 Combi-Blocks, Inc.

Ethyl (2r,4r)-1-boc-4-methylpyrrolidine-2-carboxylate

  • MDL number MFCD28053764
  • Cas Number : 1951424-81-9
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9902

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9902
Availability details for
Quantity
  • Availability
  • Add to Cart