COM497518510 Combi-Blocks, Inc.

2-(7-Methoxynaphthalen-1-yl)acetamide

  • MDL number MFCD12756807
  • Cas Number : 138113-07-2
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1313
Availability details for
Quantity
  • Availability
  • Add to Cart