COM497518575 Combi-Blocks, Inc.

(5-Chlorothiophen-3-yl)methanamine

  • MDL number MFCD19215199
  • Cas Number : 548772-41-4
  • Molecular Weight : 147.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1406

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1406
Availability details for
Quantity
  • Availability
  • Add to Cart