COM497518831 Combi-Blocks, Inc.

4-[1,2,3]Triazol-1-yl-benzaldehyde

  • MDL number MFCD07783843
  • Cas Number : 41498-10-6
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1758
Availability details for
Quantity
  • Availability
  • Add to Cart