COM497519003 Combi-Blocks, Inc.

3-(Pentan-2-yl)-1,2-oxazol-5-amine

  • MDL number MFCD10691804
  • Cas Number : 1092298-90-2
  • Molecular Weight : 154.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1990

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-1990
Availability details for
Quantity
  • Availability
  • Add to Cart