COM497519054 Combi-Blocks, Inc.

2-Methyl-1h-indole-7-carboxylic acid

  • MDL number MFCD11168318
  • Cas Number : 339366-85-7
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2050

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2050
Availability details for
Quantity
  • Availability
  • Add to Cart