COM497519099 Combi-Blocks, Inc.

4,4-Difluoro-3,3-dimethylpiperidine hydrochloride

  • MDL number MFCD11840374
  • Cas Number : 1198285-09-4
  • Molecular Weight : 185.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2105
Availability details for
Quantity
  • Availability
  • Add to Cart