COM497519140 Combi-Blocks, Inc.

tert-Butyl 2-(N'-hydroxycarbamimidoyl)pyrrolidine-1-carboxylate

  • MDL number MFCD11975692
  • Cas Number : 500024-95-3
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2149
Availability details for
Quantity
  • Availability
  • Add to Cart