COM497519220 Combi-Blocks, Inc.

3-(2-Fluoro-4-methoxyphenyl)propanoic acid

  • MDL number MFCD13659377
  • Cas Number : 852181-15-8
  • Molecular Weight : 198.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2255

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2255
Availability details for
Quantity
  • Availability
  • Add to Cart