COM497519224 Combi-Blocks, Inc.

Cis-2-methylcyclopentanol

  • MDL number MFCD13810043
  • Cas Number : 25144-05-2
  • Molecular Weight : 100.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2259

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2259
Availability details for
Quantity
  • Availability
  • Add to Cart