COM497519315 Combi-Blocks, Inc.

N-Methyl-1h-pyrazol-3-amine

  • MDL number MFCD17292026
  • Cas Number : 446866-62-2
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2369

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2369
Availability details for
Quantity
  • Availability
  • Add to Cart