COM497519466 Combi-Blocks, Inc.

Di-tert-butyl 6-oxo-1,4-diazepane-1,4-dicarboxylate

  • MDL number MFCD21362344
  • Cas Number : 459417-40-4
  • Molecular Weight : 314.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2552
Availability details for
Quantity
  • Availability
  • Add to Cart