COM497519475 Combi-Blocks, Inc.

Imidazo[1,5-a]pyridin-6-yl-methanol

  • MDL number MFCD21362365
  • Cas Number : 1225209-79-9
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2561
Availability details for
Quantity
  • Availability
  • Add to Cart