COM497519507 Combi-Blocks, Inc.

(2-Methylpyridin-3-yl)methanamine dihydrochloride

  • MDL number MFCD22209855
  • Cas Number : 58539-66-5
  • Molecular Weight : 195.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2596
Availability details for
Quantity
  • Availability
  • Add to Cart