COM497519525 Combi-Blocks, Inc.

5-(Propan-2-yl)-1,2,3-thiadiazole-4-carboxylic acid

  • MDL number MFCD22390323
  • Cas Number : 1445950-78-6
  • Molecular Weight : 172.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2616
Availability details for
Quantity
  • Availability
  • Add to Cart