COM497519708 Combi-Blocks, Inc.

3-(6-Methylpyridin-3-yl)propan-1-amine

  • MDL number MFCD13188695
  • Cas Number : 1060806-38-3
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2820

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2820
Availability details for
Quantity
  • Availability
  • Add to Cart