COM497519777 Combi-Blocks, Inc.

4-(Trifluoromethyl)azepan-4-ol hydrochloride

  • MDL number MFCD28119087
  • Cas Number : 1638764-95-0
  • Molecular Weight : 219.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2894
Availability details for
Quantity
  • Availability
  • Add to Cart