COM497520119 Combi-Blocks, Inc.

3-Hexadecanol

  • MDL number MFCD00051480
  • Cas Number : 593-03-3
  • Molecular Weight : 242.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3345

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3345
Availability details for
Quantity
  • Availability
  • Add to Cart