COM497520135 Combi-Blocks, Inc.

5-Amino-n-benzyl-2-chlorobenzamide

  • MDL number MFCD09815178
  • Cas Number : 154737-70-9
  • Molecular Weight : 260.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3362
Availability details for
Quantity
  • Availability
  • Add to Cart