COM497520176 Combi-Blocks, Inc.

Methyl 4-chloro-5,8-dimethylquinoline-2-carboxylate

  • MDL number MFCD07783794
  • Cas Number : 887589-35-7
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3403
Availability details for
Quantity
  • Availability
  • Add to Cart