COM497520332 Combi-Blocks, Inc.

2,4-Dihydro-4-[4-[4-(4-methoxyphenyl)-1-piperazinyl]phenyl]-2-(1-methylpropyl)-3h-1,2,4-triazol-3-one

  • MDL number MFCD02956607
  • Cas Number : 252964-68-4
  • Molecular Weight : 407.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3685

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3685
Availability details for
Quantity
  • Availability
  • Add to Cart