COM497520865 Combi-Blocks, Inc.

(4-Chloro-3-methoxyphenyl)methanamine

  • MDL number MFCD09752213
  • Cas Number : 247569-42-2
  • Molecular Weight : 171.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4384
Availability details for
Quantity
  • Availability
  • Add to Cart