COM497520897 Combi-Blocks, Inc.

4-Methyl-1h-indazol-6-ol

  • MDL number MFCD07781489
  • Cas Number : 885521-33-5
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4419

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4419
Availability details for
Quantity
  • Availability
  • Add to Cart