COM497520925 Combi-Blocks, Inc.

5-Bromo-4,6-dichloro-2-methylpyrimidine

  • MDL number MFCD09864954
  • Cas Number : 1086376-43-3
  • Molecular Weight : 241.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4449

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4449
Availability details for
Quantity
  • Availability
  • Add to Cart