COM497521046 Combi-Blocks, Inc.

2-Amino-4-methyl-n-phenylthiazole-5-carboxamide

  • MDL number MFCD00600635
  • Cas Number : 21452-14-2
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4584

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4584
Availability details for
Quantity
  • Availability
  • Add to Cart