COM497521282 Combi-Blocks, Inc.

1-Benzyl-6-nitro-1h-indole

  • MDL number MFCD27841266
  • Cas Number : 120627-46-5
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4864
Availability details for
Quantity
  • Availability
  • Add to Cart