COM497521308 Combi-Blocks, Inc.

4-O-Acetyl-3,6-di-o-(tert-butyldiphenylsilyl)-d-galactal

  • MDL number MFCD01863634
  • Cas Number : 342029-19-0
  • Molecular Weight : 665.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4893
Availability details for
Quantity
  • Availability
  • Add to Cart