COM497521319 Combi-Blocks, Inc.

3,6-Di-o-(tert-butyldimethylsilyl)-d-galactal

  • MDL number MFCD01863627
  • Cas Number : 111902-03-5
  • Molecular Weight : 374.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4906

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4906
Availability details for
Quantity
  • Availability
  • Add to Cart