COM497521333 Combi-Blocks, Inc.

3,4-Di-o-acetyl-6-o-(tert-butyldimethylsilyl)-d-galactal

  • MDL number MFCD01863632
  • Cas Number : 136656-49-0
  • Molecular Weight : 344.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4923

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4923
Availability details for
Quantity
  • Availability
  • Add to Cart