COM497521386 Combi-Blocks, Inc.

(2-Chloroethyl)dimethylsulfanium iodide

  • MDL number MFCD00012235
  • Cas Number : 25059-70-5
  • Molecular Weight : 252.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-5105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-5105
Availability details for
Quantity
  • Availability
  • Add to Cart