COM497521392 Combi-Blocks, Inc.

(3S)-3-Amino-3-(2-methylphenyl)propanoic acid hydrochloride

  • MDL number MFCD03427817
  • Cas Number : 490034-62-3
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-5111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-5111
Availability details for
Quantity
  • Availability
  • Add to Cart