COM497521607 Combi-Blocks, Inc.

tert-Butyl 5-amino-3-methyl-1h-indazole-1-carboxylate

  • MDL number MFCD13183005
  • Cas Number : 599183-32-1
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-5334

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-5334
Availability details for
Quantity
  • Availability
  • Add to Cart