COM497521640 Combi-Blocks, Inc.

Ethyl 2-butyl-1h-indole-6-carboxylate

  • MDL number MFCD09839183
  • Cas Number : 491601-40-2
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-5368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-5368
Availability details for
Quantity
  • Availability
  • Add to Cart