COM497522277 Combi-Blocks, Inc.

5-Methoxyquinoline-3-carboxylic acid

  • MDL number MFCD21607487
  • Cas Number : 1361091-98-6
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-6077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-6077
Availability details for
Quantity
  • Availability
  • Add to Cart