COM497522442 Combi-Blocks, Inc.

8-(Trifluoromethyl)quinolin-5-amine

  • MDL number MFCD22627921
  • Cas Number : 161431-57-8
  • Molecular Weight : 212.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7542
Availability details for
Quantity
  • Availability
  • Add to Cart