COM497522673 Combi-Blocks, Inc.

tert-Butyl n-[(1-aminocyclopentyl)methyl]carbamate

  • MDL number MFCD18483181
  • Cas Number : 1311318-21-4
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-8647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-8647
Availability details for
Quantity
  • Availability
  • Add to Cart