COM497522760 Combi-Blocks, Inc.

2-(4-Bromophenyl)-3-hydroxypropanenitrile

  • MDL number MFCD28405198
  • Cas Number : 123654-61-5
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-9730

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-9730
Availability details for
Quantity
  • Availability
  • Add to Cart