COM497522787 Combi-Blocks, Inc.

2-(Pyrrolidin-1-yl)benzoic acid hydrochloride

  • MDL number MFCD28962946
  • Cas Number : 1855911-27-1
  • Molecular Weight : 227.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4296

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4296
Availability details for
Quantity
  • Availability
  • Add to Cart