COM497522918 Combi-Blocks, Inc.

3-Amino-4-cyclopropylthiophene-2-carbonitrile hcl

  • MDL number MFCD28133915
  • Cas Number : 1603054-97-2
  • Molecular Weight : 200.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1579
Availability details for
Quantity
  • Availability
  • Add to Cart