COM497522938 Combi-Blocks, Inc.

Dimethyl 5-formylisophthalate

  • MDL number MFCD16987722
  • Cas Number : 164073-80-7
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1899
Availability details for
Quantity
  • Availability
  • Add to Cart