COM497523060 Combi-Blocks, Inc.

8-Chloro-3-(propan-2-yl)-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD20303915
  • Cas Number : 140910-91-4
  • Molecular Weight : 196.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3166
Availability details for
Quantity
  • Availability
  • Add to Cart