COM497523101 Combi-Blocks, Inc.

tert-Butyl 3-cyano-1H-indole-1-carboxylate

  • MDL number MFCD16037144
  • Cas Number : 908244-43-9
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3223

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3223
Availability details for
Quantity
  • Availability
  • Add to Cart