COM497523102 Combi-Blocks, Inc.

Ditert-butyl cyclohexane-1,3-diyldicarbamate

  • MDL number MFCD18252218
  • Cas Number : 1823244-92-3
  • Molecular Weight : 314.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3224
Availability details for
Quantity
  • Availability
  • Add to Cart