COM497523141 Combi-Blocks, Inc.

4,4-Dimethyl-2-(2-(trifluoromethyl) phenyl)-4,5-dihydrooxazole

  • MDL number MFCD18206200
  • Cas Number : 263382-53-2
  • Molecular Weight : 243.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3266
Availability details for
Quantity
  • Availability
  • Add to Cart