COM497523164 Combi-Blocks, Inc.

Methyl 4-amino-1-methylpiperidine-4-carboxylate hydrochloride

  • MDL number MFCD18206236
  • Cas Number : 161909-44-0
  • Molecular Weight : 208.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3289

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3289
Availability details for
Quantity
  • Availability
  • Add to Cart