COM497523168 Combi-Blocks, Inc.

Methyl 3-bromo-4-(methylamino)benzoate

  • MDL number MFCD11110863
  • Cas Number : 1131615-02-5
  • Molecular Weight : 244.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3293

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3293
Availability details for
Quantity
  • Availability
  • Add to Cart