COM497523185 Combi-Blocks, Inc.

Octa-3,5-diyne-1,8-diol

  • MDL number MFCD00190210
  • Cas Number : 15808-23-8
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3310
Availability details for
Quantity
  • Availability
  • Add to Cart