COM497523223 Combi-Blocks, Inc.

1-Acetylpiperidine-4-carboxamide

  • MDL number MFCD02029734
  • Cas Number : 530120-27-5
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3348

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3348
Availability details for
Quantity
  • Availability
  • Add to Cart