COM497523228 Combi-Blocks, Inc.

N-[2-[3-(Aminosulfonyl)-4-methoxyphenyl]ethyl]-5-chloro-2-methoxy-benzamide

  • MDL number MFCD16620222
  • Cas Number : 158751-78-1
  • Molecular Weight : 398.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3353
Availability details for
Quantity
  • Availability
  • Add to Cart