COM497523231 Combi-Blocks, Inc.

2-(2H-1,2,3-Benzotriazol-2-yl)ethan-1-ol

  • MDL number MFCD00835235
  • Cas Number : 939-72-0
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3357

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3357
Availability details for
Quantity
  • Availability
  • Add to Cart